accessibility menu, dialog, popup

UserName

Viewing profile as user:
NONH2

CAS RN 86556-09-4

IUPAC Name: 3-(pyridin-2-yloxy)aniline
Synonyms: 3-(pyridin-2-yloxy)aniline
Molecular Weight (g/mol): 186.21
Molecular Formula: C11H10N2O
InChi Key: CLQMOPKXIBQKKG-UHFFFAOYSA-N
SMILES: NC1=CC(OC2=CC=CC=N2)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results