accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOHHOCH3OOHHOH2OH2OH2OH2OH2OCa

CAS RN 865717-20-0

IUPAC Name: bis(2-hydroxypropanoic acid) calcium pentahydrate
Synonyms: bis(lactic acid) calcium pentahydrate
Molecular Weight (g/mol): 310.31
Molecular Formula: C6H22CaO11
InChi Key: YNIXORYWSZWLSA-UHFFFAOYNA-N
SMILES: O.O.O.O.O.[Ca].CC(O)C(O)=O.CC(O)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results