Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium L-lactate pentahydrate, 98%

Catalog No. AC250200010
Change view
Click to view available options
Quantity:
1 kg
Packaging:
Plastic bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC250200010 1 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC250200010 Supplier Thermo Scientific Chemicals Supplier No. 250200010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 865717-20-0 | C6H10CaO6·5H2O | 308.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 865717-20-0 , 28305-25-1
Molecular Formula C6H10CaO6·5H2O
Molecular Weight (g/mol) 308.29
MDL Number MFCD00287281
InChI Key YNIXORYWSZWLSA-VFGZHDCSSA-N
Synonym Lactic acid, calcium salt pentahydrate, (S)-2-Hydroxypropanoic acid, calcium salt pentahydrate
PubChem CID 131886284
IUPAC Name calcium;(2S)-2-hydroxypropanoic acid;pentahydrate
SMILES CC(C(=O)O)O.CC(C(=O)O)O.O.O.O.O.O.[Ca]

Specifications

Melting Point 240.0°C
Color White
pH 6.0 to 8.0 (5% aq. soln.)
Physical Form Powder
Quantity 1 kg
Assay Percent Range 97.5% min. (on dry substance) (Complexometry)
Beilstein 03, IV, 636
Merck Index 14, 1678
Solubility Information Solubility in water: 90g/L (25°C). Other solubilities: practically insoluble in alcohol
Formula Weight 308.29
Percent Purity 98%
Infrared Spectrum Authentic
Loss on Drying 22.0% to 27.0%
Packaging Plastic bottle
Chemical Name or Material Calcium L-lactate pentahydrate, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.