CAS RN 86604-78-6
IUPAC Name:
(4-methoxy-3,5-dimethylpyridin-2-yl)methanol
Synonyms:
(4-methoxy-3,5-dimethylpyridin-2-yl)methanol
Molecular Weight (g/mol):
167.21
Molecular Formula:
C9H13NO2
InChi Key:
PSEPRWKZZJWRCB-UHFFFAOYSA-N
SMILES:
COC1=C(C)C=NC(CO)=C1C
1
–
1
of
1
results
1
4-Methoxy-3,5-dimethyl-2-pyridinemethanol 98.0+%, TCI America™
CAS: 86604-78-6 Molecular Formula: C9H13NO2 Molecular Weight (g/mol): 167.21 MDL Number: MFCD01076195 InChI Key: PSEPRWKZZJWRCB-UHFFFAOYSA-N Synonym: 2-Hydroxymethyl-4-methoxy-3,5-dimethylpyridine PubChem CID: 737823 IUPAC Name: (4-methoxy-3,5-dimethylpyridin-2-yl)methanol SMILES: COC1=C(C)C=NC(CO)=C1C