accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 867-13-0

IUPAC Name: ethyl 2-(diethoxyphosphoryl)acetate
Synonyms: triethyl phosphonoacetate
Molecular Weight (g/mol): 224.19
Molecular Formula: C8H17O5P
InChi Key: GGUBFICZYGKNTD-UHFFFAOYSA-N
SMILES: CCOC(=O)CP(=O)(OCC)OCC
Boiling Point 
  • (3)
  • (3)
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (3)
  • (6)
Quantity 
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (3)
  • (6)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (6)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (6)