Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Triethyl Phosphonoacetate

Catalog No. T6130125G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T6130125G 25 g
T61301100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. T6130125G Supplier Sigma Aldrich Supplier No. T6130125G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reactant involved in:Horner-Wadsworth-Emmons reactionsIntramolecular Heck-type cyclization and isomerizationsTsuji-Trost type reactionsIntramolecular aryne-ene reactionsSynthesis of aldehydes

Specifications

CAS 867-13-0
Density 1.13 g/mL at 25°C
Boiling Point 142°C to 145°C (9 mmHg, lit.)
Molecular Formula C8H17O5P
Refractive Index n20/D 1.431
Linear Formula (C2H5O)2P(O)CH2CO2C2H5
MDL Number MFCD00009177
Quantity 25 g
Synonym (Diethoxyphosphinyl)acetic acid ethyl ester; Diethyl ethoxycarbonylmethylphosphonate; NSC 13898; NSC 16128; Triethyl carboxymethylphosphonate
Molecular Weight (g/mol) 224.19 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.