accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaH3COHOO

CAS RN 867-56-1

IUPAC Name: sodium 2-hydroxypropanoate
Synonyms: sodium lactate
Molecular Weight (g/mol): 112.06
Molecular Formula: C3H5NaO3
InChi Key: NGSFWBMYFKHRBD-UHFFFAOYNA-M
SMILES: [Na+].CC(O)C([O-])=O
Molecular Weight (g/mol) 
  • (6)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
Grade 
  • (1)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (3)
  • (6)
  • (2)
  • (6)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)