Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Lactic Acid, Sodium Salt, 60 wt% Solution in Water

Catalog No. AC250300050
Change view
Click to view available options
Quantity:
5 L
Packaging:
Plastic drum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC250300050 5 L Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC250300050 Supplier Thermo Scientific Chemicals Supplier No. 250300050
Add to Cart

CAS: 867-56-1 | C3H5NaO3 | 112.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material L-Lactic Acid, Sodium Salt
Name Note 60 wt% Solution in Water
CAS 867-56-1, 64-17-5
Density 1.3300g/mL
Packaging Plastic drum
Molecular Formula C3H5NaO3
Refractive Index 1.4220 to 1.4250
Linear Formula CH3CH(OH)CO2Na
Quantity 5 L
MDL Number MFCD00066576
Specific Gravity 1.33
Merck Index 14,8635
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol
Molecular Weight (g/mol) 112.06
Viscosity 80-160 mPa.s (20°C)
Formula Weight 112.06
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.