accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaH3CH3COHOHONHOO

CAS RN 867-81-2

IUPAC Name: sodium 3-(2,4-dihydroxy-4-methylpentanamido)propanoate
Synonyms: sodium pantothenate
Molecular Weight (g/mol): 241.22
Molecular Formula: C9H16NNaO5
InChi Key: ROMBWECPDOCQMZ-UHFFFAOYNA-M
SMILES: [Na+].CC(C)(O)CC(O)C(=O)NCCC([O-])=O
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Grade 
  • (3)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)