Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Pantothenic Acid Sodium Salt, Spectrum™ Chemical
SDP

Catalog No. 18602348 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-348 1 kg Amber Glass Bottle
18-602-346 25 g Amber Glass Bottle
18-602-347 100 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-602-348 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. P14681KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

P1468, 867-81-2, C9H16NO5Na

Spectrum™ Chemical D-Pantothenic Acid Sodium Salt is also known as pantothenate and is soluble in water. It is considered an essential nutrient for several types of animals. It is also an ingredient in some skin or hair products. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 867-81-2
Molecular Formula C9H16NNaO5
Molecular Weight (g/mol) 241.22
MDL Number MFCD00002767
InChI Key ROMBWECPDOCQMZ-UHFFFAOYNA-M
IUPAC Name sodium 3-(2,4-dihydroxy-4-methylpentanamido)propanoate
SMILES [Na+].CC(C)(O)CC(O)C(=O)NCCC([O-])=O

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 1 kg
Grade Technical
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.