CAS RN 868-54-2
IUPAC Name:
2-aminoprop-1-ene-1,1,3-tricarbonitrile
Synonyms:
tricyanoaminopropene
Molecular Weight (g/mol):
132.13
Molecular Formula:
C6H4N4
InChi Key:
BNHGNFYPZNDLAF-UHFFFAOYSA-N
SMILES:
NC(CC#N)=C(C#N)C#N