Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-1,1,3-tricyano-1-propene 98.0+%, TCI America™
SDP

Catalog No. A109725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A109725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A109725G Supplier TCI America Supplier No. A109725G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 868-54-2
Melting Point 169°C
Molecular Formula C6H4N4
Quantity 25 g
MDL Number MFCD00001851
UN Number 3439
Synonym malononitrile dimer, 2-amino-1-propene-1,1,3-tricarbonitrile, 2-amino-1,1,3-tricyano-1-propene, 2-amino-1,1,3-tricyanopropene, upjohn 9189, tcap, 2-amino-1,1,3-propenetricarbonitrile, 1,1,3-tricyano-2-aminopropene, 1-amino-2-cyanoethylidene malononitrile, tcap van
InChI Key BNHGNFYPZNDLAF-UHFFFAOYSA-N
SMILES C(C#N)C(=C(C#N)C#N)N
IUPAC Name 2-aminoprop-1-ene-1,1,3-tricarbonitrile
Molecular Weight (g/mol) 132.126
PubChem CID 13356
Formula Weight 132.13
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-1,1,3-tricyano-1-propene
Show More Show Less
RTECSNumber UD2340000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.