accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 868735-43-7

IUPAC Name: tert-butyl N-(4-bromo-3-fluorophenyl)carbamate
Synonyms: tert-butyl N-(4-bromo-3-fluorophenyl)carbamate
Molecular Weight (g/mol): 290.13
Molecular Formula: C11H13BrFNO2
InChi Key: FQLYUJLJZFATNI-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NC1=CC(F)=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results