Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Boc-4-bromo-3-fluoroaniline, 96%, Thermo Scientific Chemicals

Catalog No. AAH5939803
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5939803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5939803 Supplier Thermo Scientific Chemicals Supplier No. H5939803
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 868735-43-7 | Molecular Formula: C11H13BrFNO2 | Molecular Weight: 290.132 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 868735-43-7
Molecular Formula C11H13BrFNO2
Molecular Weight (g/mol) 290.132
MDL Number MFCD11976284
InChI Key FQLYUJLJZFATNI-UHFFFAOYSA-N
Synonym tert-butyl 4-bromo-3-fluorophenyl carbamate, tert-butyl n-4-bromo-3-fluorophenyl carbamate, tert-butyl 4-bromo-3-fluorophenylcarbamate, 4-bromo-3-fluoro-phenyl-carbamic acid tert-butyl ester, n-boc-4-bromo-3-fluoroaniline
PubChem CID 59852943
IUPAC Name tert-butyl N-(4-bromo-3-fluorophenyl)carbamate
SMILES CC(C)(C)OC(=O)NC1=CC(=C(C=C1)Br)F

Specifications

Melting Point 68°C to 70°C
Quantity 1 g
UN Number UN2811
Beilstein 14460051
Formula Weight 290.13
Percent Purity 96%
Chemical Name or Material N-Boc-4-bromo-3-fluoroaniline
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.