CAS RN 87-10-5
CAS RN 87-10-5
IUPAC Name:
3,5-dibromo-N-(4-bromophenyl)-2-hydroxybenzamide
Synonyms:
tribromosalicylanilide tribromsalanum caswell no. 863 tribromsalan asc-4 vancide tbs TBS temasept IV temasept II trisanil
Molecular Weight (g/mol):
449.92
Molecular Formula:
C13H8Br3NO2
InChi Key:
KVSKGMLNBAPGKH-UHFFFAOYSA-N
SMILES:
OC1=C(Br)C=C(Br)C=C1C(=O)NC1=CC=C(Br)C=C1
1
–
1
of
1
results
1
3,5,4'-Tribromosalicylanilide 98.0+%, TCI America™

CAS: 87-10-5 Molecular Formula: C13H8Br3NO2 Molecular Weight (g/mol): 449.92 MDL Number: MFCD00045740 InChI Key: KVSKGMLNBAPGKH-UHFFFAOYSA-N PubChem CID: 14868 ChEBI: CHEBI:127105 IUPAC Name: 3,5-dibromo-N-(4-bromophenyl)-2-hydroxybenzamide SMILES: OC1=C(Br)C=C(Br)C=C1C(=O)NC1=CC=C(Br)C=C1