Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5,4'-Tribromosalicylanilide 98.0+%, TCI America™
SDP

Catalog No. T035625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T035625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. T035625G Supplier TCI America Supplier No. T035625G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 87-10-5
Molecular Formula C13H8Br3NO2
Molecular Weight (g/mol) 449.92
MDL Number MFCD00045740
InChI Key KVSKGMLNBAPGKH-UHFFFAOYSA-N
PubChem CID 14868
ChEBI CHEBI:127105
IUPAC Name 3,5-dibromo-N-(4-bromophenyl)-2-hydroxybenzamide
SMILES OC1=C(Br)C=C(Br)C=C1C(=O)NC1=CC=C(Br)C=C1

Specifications

Melting Point 225°C
Quantity 25 g
Formula Weight 449.92
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3,5,4′-Tribromosalicylanilide
EINECSNumber (3)-1556&(3)-1573&(9)-0911
RTECSNumber VN8925000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.