accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOOH3COOCH3

CAS RN 87-13-8

IUPAC Name: 1,3-diethyl 2-(ethoxymethylidene)propanedioate
Synonyms: 1,3-diethyl 2-(ethoxymethylidene)propanedioate
Molecular Weight (g/mol): 216.23
Molecular Formula: C10H16O5
InChi Key: LTMHNWPUDSTBKD-UHFFFAOYSA-N
SMILES: CCOC=C(C(=O)OCC)C(=O)OCC
Boiling Point 
  • (4)
  • (3)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (4)
  • (7)
Percent Purity 
  • (4)
  • (3)
  • (2)
  • (2)
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (3)
  • (6)
  • (4)
  • (3)
  • (2)
  • (2)
  • (4)
  • (7)
  • (4)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (5)