Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl ethoxymethylenemalonate, 98+%

Catalog No. AAA1377636
Change view
Click to view available options
Quantity:
50 g
100 g
250 g
500 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1377636 500 g
AAA1377618 50 g
AAA1377622 100 g
AAA1377630 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAA1377636 Supplier Thermo Scientific Chemicals Supplier No. A1377636
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 87-13-8 | C10H16O5 | 216.233 g/mol

Diethyl ethoxymethylenemalonate is used for synthesis of pyrido[3,2-e]pyrimido[1,2-c]pyrimidines. Initially, it was used to monitor lysine decarboxylase activity. It is useful in the determination of amino acids by precolumn derivatization by using reversed-phase high-performance liquid chromatography (HPLC). It plays an important role in the Gould-Jacobs reaction to prepare quinolines. For example, anile reacts with this reagent to give 4-hydroxyquinoline. It acts as an intermediate in the production of flumequine, which is a fluoroquinolone antibiotic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl ethoxymethylenemalonate is used for synthesis of pyrido[3,2-e]pyrimido[1,2-c]pyrimidines. Initially, it was used to monitor lysine decarboxylase activity. It is useful in the determination of amino acids by precolumn derivatization by using reversed-phase high-performance liquid chromatography (HPLC). It plays an important role in the Gould-Jacobs reaction to prepare quinolines. For example, anile reacts with this reagent to give 4-hydroxyquinoline. It acts as an intermediate in the production of flumequine, which is a fluoroquinolone antibiotic.

Solubility
Miscible with chloroform, dichloromethane and methanol. Immiscible with water.

Notes
Incompatible with acids, bases, oxidizing agents and reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87-13-8
Molecular Formula C10H16O5
Molecular Weight (g/mol) 216.233
MDL Number MFCD00009148
InChI Key LTMHNWPUDSTBKD-UHFFFAOYSA-N
Synonym diethyl ethoxymethylenemalonate, diethyl 2-ethoxymethylene malonate, ethoxymethylenemalonic acid diethyl ester, diethyl ethoxymethylene malonate, diethyl emme, propanedioic acid, ethoxymethylene-, diethyl ester, diethylethoxymethylenemalonate, 1,3-diethyl 2-ethoxymethylidene propanedioate, diethyl ethoxymethylidene propanedioate
PubChem CID 6871
IUPAC Name diethyl 2-(ethoxymethylidene)propanedioate
SMILES CCOC=C(C(=O)OCC)C(=O)OCC

Specifications

Melting Point -33°C
Density 1.08
Boiling Point 136°C to 139°C (10 mmHg)
Flash Point 144°C (291°F)
Odor Ester-like
Refractive Index 1.462
Quantity 500 g
Beilstein 880058
Solubility Information Miscible with chloroform,dichloromethane and methanol. Immiscible with water.
Formula Weight 216.23
Percent Purity ≥98%
Chemical Name or Material Diethyl ethoxymethylenemalonate
Show More Show Less
Hazard Category H302-H314-H317-H334-H335
Hazard Statement GHS H Statement
H334-H315-H319-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Precautionary Statement P260-P264b-P270-P271-P272-P280-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c
EINECSNumber 201-725-7
RTECSNumber OO1100000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.