accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 87-42-3

IUPAC Name: 6-chloro-7H-purine
Synonyms: 6-chloropurine
Molecular Weight (g/mol): 154.56
Molecular Formula: C5H3ClN4
InChi Key: ZKBQDFAWXLTYKS-UHFFFAOYSA-N
SMILES: ClC1=C2NC=NC2=NC=N1
Molecular Weight (g/mol) 
  • (5)
  • (4)
Percent Purity 
  • (2)
  • (1)
  • (4)
  • (2)
Quantity 
  • (2)
  • (3)
  • (1)
  • (3)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (5)
  • (5)
  • (4)
  • (2)
  • (1)
  • (4)
  • (2)
  • (2)
  • (3)
  • (1)
  • (3)
  • (4)