CAS RN 87-68-3
IUPAC Name:
hexachlorobuta-1,3-diene
Synonyms:
hexachlorobutadiene
Molecular Weight (g/mol):
260.74
Molecular Formula:
C4Cl6
InChi Key:
RWNKSTSCBHKHTB-UHFFFAOYSA-N
SMILES:
ClC(Cl)=C(Cl)C(Cl)=C(Cl)Cl
1
–
1
of
1
results
1
Hexachlorobenzene Reference Material, 1,000 μg/mL In Acetone, 1 mL
CAS: 87-68-3 Molecular Formula: C4Cl6 Molecular Weight (g/mol): 260.744 InChI Key: RWNKSTSCBHKHTB-UHFFFAOYSA-N PubChem CID: 6901 ChEBI: CHEBI:5691 IUPAC Name: 1,1,2,3,4,4-hexachlorobuta-1,3-diene SMILES: C(=C(Cl)Cl)(C(=C(Cl)Cl)Cl)Cl