Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexachlorobenzene Reference Material, 1,000 μg/mL In Acetone, 1 mL

Catalog No. USEPA1125
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-EPA-1125 1 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. US-EPA-1125 Supplier Agilent Technologies Supplier No. EPA11251
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 87-68-3 , 67-64-1
Molecular Formula C4Cl6
Molecular Weight (g/mol) 260.744
InChI Key RWNKSTSCBHKHTB-UHFFFAOYSA-N
PubChem CID 6901
ChEBI CHEBI:5691
IUPAC Name 1,1,2,3,4,4-hexachlorobuta-1,3-diene
SMILES C(=C(Cl)Cl)(C(=C(Cl)Cl)Cl)Cl

Specifications

CAS Max % 99.87, 0.12
Analyte or Component Names methanol,fluoranthene
Element Methanol, Fluoranthene
Solvent or Matrix Methanol
Quantity 1 x 1 mL
Concentration or Composition (by Analyte or Components) 5000μg/mL
Chemical Name or Material Hexachlorobenzene Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.