accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 870-50-8

IUPAC Name: N-{[(tert-butoxy)carbonyl]imino}(tert-butoxy)formamide
Synonyms: N-[(tert-butoxycarbonyl)imino](tert-butoxy)formamide
Molecular Weight (g/mol): 230.26
Molecular Formula: C10H18N2O4
InChi Key: QKSQWQOAUQFORH-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N=NC(=O)OC(C)(C)C
Molecular Weight (g/mol) 
  • (3)
  • (4)
  • (3)
Quantity 
  • (1)
  • (5)
  • (4)
Percent Purity 
  • (3)
  • (2)
Physical Form 
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (4)
  • (1)
  • (5)
  • (3)
  • (4)
  • (3)
  • (1)
  • (5)
  • (4)
  • (3)
  • (2)
  • (3)
  • (1)