Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Di-tert-butyl Azodicarboxylate (20% in Toluene), TCI America™
SDP

Catalog No. D354425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D354425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D354425G Supplier TCI America Supplier No. D354425G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 870-50-8
Molecular Formula C10H18N2O4
Molecular Weight (g/mol) 230.264
MDL Number MFCD00015001
InChI Key QKSQWQOAUQFORH-VAWYXSNFSA-N
Synonym di-tert-butyl azodicarboxylate, di-tert-butylazodicarboxylate, di-tert-butyl diazene-1,2-dicarboxylate, azodicarboxylic acid di-tert-butyl ester, di-tert-butyl azodicarboxylate dbad, pubchem17548, n-tert-butoxycarbonyl imino tert-butoxy formamide, tert-butyl tert-butyl oxycarbonyl diazenyl formate, n-tert-butoxy carbonyl imino tert-butoxy formamide, 1,2-diazenedicarboxylic acid, 1,2-bis 1,1-dimethylethyl ester
PubChem CID 6034084
IUPAC Name tert-butyl (NE)-N-[(2-methylpropan-2-yl)oxycarbonylimino]carbamate
SMILES CC(C)(C)OC(=O)N=NC(=O)OC(C)(C)C

Specifications

Quantity 25 g
UN Number 1993
Formula Weight 230.26
Physical Form Liquid
Chemical Name or Material Di-tert-butyl Azodicarboxylate (20% in Toluene)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.