accessibility menu, dialog, popup

UserName

Viewing profile as user:
HNNNNNClClON

CAS RN 870061-27-1

IUPAC Name: 1-(2,3-dichlorophenyl)-N-{[2-(pyridin-2-yloxy)phenyl]methyl}-1H-1,2,3,4-tetrazol-5-amine
Synonyms: 1-(2,3-dichlorophenyl)-N-{[2-(pyridin-2-yloxy)phenyl]methyl}-1,2,3,4-tetrazol-5-amine
Molecular Weight (g/mol): 413.26
Molecular Formula: C19H14Cl2N6O
InChi Key: GMVNBKZQJFRFAR-UHFFFAOYSA-N
SMILES: ClC1=CC=CC(N2N=NN=C2NCC2=CC=CC=C2OC2=CC=CC=N2)=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results