accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 870076-72-5

IUPAC Name: 4-[2-(tert-butylamino)-1-methoxyethyl]-2-(hydroxymethyl)phenol
Synonyms: 4-[2-(tert-butylamino)-1-methoxyethyl]-2-(hydroxymethyl)phenol
Molecular Weight (g/mol): 253.34
Molecular Formula: C14H23NO3
InChi Key: UMHASVFLCHGDPW-UHFFFAOYNA-N
SMILES: COC(CNC(C)(C)C)C1=CC(CO)=C(O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results