Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-[2-(tert-Butylamino)-1-methoxyethyl]-2-(hydroxymethyl)phenol, MilliporeSigma™ Supelco™

Catalog No. 111022809 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
11-102-2809 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-2809 Supplier MilliporeSigma Supelco Supplier No. 9280450MG

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 870076-72-5
Molecular Formula C14H23NO3
Molecular Weight (g/mol) 253.34
MDL Number MFCD23160660
InChI Key UMHASVFLCHGDPW-UHFFFAOYNA-N
Synonym Salbutamol impurity A (PhEur); 5-[(1 RS)-2-[(1,1-Dimethylethyl)amino]-1-methoxyethyl]-2-hydroxyphenyl]methanol; 5-[2-[(1,1-Dimethylethyl)amino]-1-methoxyethyl]-2-hydroxybenzenemethanol
IUPAC Name 4-[2-(tert-butylamino)-1-methoxyethyl]-2-(hydroxymethyl)phenol
SMILES COC(CNC(C)(C)C)C1=CC(CO)=C(O)C=C1

Specifications

Quantity 50 mg
Linear Formula C14H23NO3
UN Number NONH for all modes of transport
Formula Weight 253.34
Percent Purity ≥98% (HPLC)
ShelfLife Limited shelf life, expiry date on the label
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.