accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 87016-67-9

IUPAC Name: 11-bromo-2,5,8,15,18,21-hexaoxatricyclo[20.4.0.0⁹,¹⁴]hexacosa-1(26),9,11,13,22,24-hexaene
Synonyms: 11-bromo-2,5,8,15,18,21-hexaoxatricyclo[20.4.0.0⁹,¹⁴]hexacosa-1(26),9,11,13,22,24-hexaene
Molecular Weight (g/mol): 439.3
Molecular Formula: C20H23BrO6
InChi Key: SDMPHKFSDSMFIL-UHFFFAOYSA-N
SMILES: BrC1=CC=C2OCCOCCOC3=CC=CC=C3OCCOCCOC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results