CAS RN 870521-31-6
CAS RN 870521-31-6
IUPAC Name:
5-bromo-2-(propan-2-yloxy)pyridine
Synonyms:
5-bromo-2-isopropoxypyridine
Molecular Weight (g/mol):
216.08
Molecular Formula:
C8H10BrNO
InChi Key:
FSZRSTNRZUIEMA-UHFFFAOYSA-N
SMILES:
CC(C)OC1=NC=C(Br)C=C1
1
–
1
of
1
results
1
Thermo Scientific Chemicals 5-Bromo-2-isopropoxypyridine, 97%
CAS: 870521-31-6 Molecular Formula: C8H10BrNO Molecular Weight (g/mol): 216.08 MDL Number: MFCD07368869 InChI Key: FSZRSTNRZUIEMA-UHFFFAOYSA-N Synonym: 5-bromo-2-isopropoxypyridine,5-bromo-2-propan-2-yloxy pyridine,5-bromo-2-isopropoxy-pyridine,3-bromo-6-i-propoxy pyridine,pyridine, 5-bromo-2-1-methylethoxy,pubchem22060,acmc-209qd4,2-isopropoxy-5-bromo-pyridine,5-bromo-2-iso-propoxypyridine,5-bromo-2-isopropyloxypyridine PubChem CID: 21935068 IUPAC Name: 5-bromo-2-propan-2-yloxypyridine SMILES: CC(C)OC1=NC=C(Br)C=C1