accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 870778-89-5

IUPAC Name: (2,4-dibutoxyphenyl)boronic acid
Synonyms: 2,4-dibutoxyphenylboronic acid
Molecular Weight (g/mol): 266.14
Molecular Formula: C14H23BO4
InChi Key: RZGMDQYKUFKOMM-UHFFFAOYSA-N
SMILES: CCCCOC1=CC(OCCCC)=C(C=C1)B(O)O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results