Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dibutoxyphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. D42635G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4263-5G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D4263-5G Supplier TCI America Supplier No. D42635G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 870778-89-5
Molecular Formula C14H23BO4
Molecular Weight (g/mol) 266.14
MDL Number MFCD07784348
InChI Key RZGMDQYKUFKOMM-UHFFFAOYSA-N
Synonym 2,4-Dibutoxybenzeneboronic Acid
PubChem CID 16217895
IUPAC Name (2,4-dibutoxyphenyl)boronic acid
SMILES CCCCOC1=CC(OCCCC)=C(C=C1)B(O)O

Specifications

Melting Point 104°C
Quantity 5 g
Formula Weight 266.14
Physical Form Crystalline Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.