accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 870779-02-5

IUPAC Name: (2,6-difluoro-3-methoxyphenyl)boronic acid
Synonyms: 2,6-difluoro-3-methoxyphenylboronic acid
Molecular Weight (g/mol): 187.94
Molecular Formula: C7H7BF2O3
InChi Key: WSRQWTCBDHWVGY-UHFFFAOYSA-N
SMILES: COC1=C(F)C(B(O)O)=C(F)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results