Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Difluoro-3-methoxyphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. D50065G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D5006-5G 5 g
D50061G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D5006-5G Supplier TCI America Supplier No. D50065G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 870779-02-5
Molecular Formula C7H7BF2O3
MDL Number MFCD06657880
InChI Key WSRQWTCBDHWVGY-UHFFFAOYSA-N
Synonym 2,6-Difluoro-3-methoxybenzeneboronic Acid
PubChem CID 2783131
IUPAC Name (2,6-difluoro-3-methoxyphenyl)boronic acid

Specifications

Melting Point 114°C
Color White-Yellow
Quantity 5 g
SMILES COC1=C(F)C(B(O)O)=C(F)C=C1
Molecular Weight (g/mol) 187.94
Formula Weight 187.94
Physical Form Crystalline Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.