accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 870991-70-1

IUPAC Name: 2-[1,2-diamino-2-(2-hydroxyphenyl)ethyl]phenol
Synonyms: 2-[1,2-diamino-2-(2-hydroxyphenyl)ethyl]phenol
Molecular Weight (g/mol): 244.29
Molecular Formula: C14H16N2O2
InChi Key: MRNPLGLZBUDMRE-UHFFFAOYNA-N
SMILES: NC(C(N)C1=CC=CC=C1O)C1=CC=CC=C1O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results