Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1R,2R)-1,2-Bis(2-hydroxyphenyl)ethylenediamine 95.0+%, TCI America™
SDP

Catalog No. B30121G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B30121G 1 g
B3012100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B30121G Supplier TCI America Supplier No. B30121G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 870991-70-1
Molecular Formula C14H16N2O2
Molecular Weight (g/mol) 244.294
MDL Number MFCD09751760
InChI Key MRNPLGLZBUDMRE-ZIAGYGMSSA-N
PubChem CID 16218403
IUPAC Name 2-[(1R,2R)-1,2-diamino-2-(2-hydroxyphenyl)ethyl]phenol
SMILES C1=CC=C(C(=C1)C(C(C2=CC=CC=C2O)N)N)O

Specifications

Melting Point 158°C
Quantity 1 g
Synonym (1R,2R)-1,2-Diamino-1,2-bis(2-hydroxyphenyl)ethane, 2,2′C-[(1R,2R)-1,2-Diaminoethylene]bisphenol
Formula Weight 244.29
Percent Purity ≥95.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (1R,2R)-1,2-Bis(2-hydroxyphenyl)ethylenediamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.