accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NSNH2

CAS RN 871-76-1

IUPAC Name: 2-[(2-aminoethyl)sulfanyl]ethan-1-amine
Synonyms: 2-[(2-aminoethyl)sulfanyl]ethanamine
Molecular Weight (g/mol): 120.21
Molecular Formula: C4H12N2S
InChi Key: JONTXEXBTWSUKE-UHFFFAOYSA-N
SMILES: NCCSCCN

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results