Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2'-Thiobis(ethylamine) 98.0+%, TCI America™
SDP

Catalog No. T153810G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T153810G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T153810G Supplier TCI America Supplier No. T153810G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 871-76-1
Molecular Formula C4H12N2S
Molecular Weight (g/mol) 120.214
MDL Number MFCD00039783
InChI Key JONTXEXBTWSUKE-UHFFFAOYSA-N
Synonym 2-Aminoethyl Sulfide, Bis(2-aminoethyl) Sulfide, 2,2′C-Diaminodiethyl Sulfide
PubChem CID 70096
IUPAC Name 2-(2-aminoethylsulfanyl)ethanamine
SMILES C(CSCCN)N

Specifications

Color Yellow
Boiling Point 120°C
Quantity 10 g
UN Number 2735
Formula Weight 120.21
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material 2,2′-Thiobis(ethylamine)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.