accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOO

CAS RN 87121-89-9

IUPAC Name: ethyl 3-oxocyclobutane-1-carboxylate
Synonyms: ethyl 3-oxocyclobutane-1-carboxylate
Molecular Weight (g/mol): 142.15
Molecular Formula: C7H10O3
InChi Key: BXBRFSMPBOTZHJ-UHFFFAOYSA-N
SMILES: CCOC(=O)C1CC(=O)C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results