Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 3-oxocyclobutanecarboxylate, 97%, Thermo Scientific™

Catalog No. AAH6242006
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6242006 5 g
AAH6242003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6242006 Supplier Thermo Scientific Chemicals Supplier No. H6242006

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Ethyl 3-oxocyclobutanecarboxylate is used as pharmaceutical intermediate.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87121-89-9
Molecular Formula C7H10O3
Molecular Weight (g/mol) 142.154
MDL Number MFCD09834114
InChI Key BXBRFSMPBOTZHJ-UHFFFAOYSA-N
PubChem CID 22467070
IUPAC Name ethyl 3-oxocyclobutane-1-carboxylate
SMILES CCOC(=O)C1CC(=O)C1

Specifications

Quantity 5 g
Synonym ethyl 3-oxocyclobutanecarboxylate, 3-oxo-cyclobutanecarboxylic acid ethyl ester, 3-oxocyclobutanecarboxylic acid ethyl ester, cyclobutanecarboxylic acid, 3-oxo-, ethyl ester, 3-ethoxycarbonylcyclobutanone, ethoxycarbonyl-3-cyclobutanone, ethyl3-oxocyclobutanecarboxylate, 3-ethoxycarbonyl-1-cyclobutanone, ethyl 3-oxo-cyclobutanecarboxylate
Formula Weight 142.15
Percent Purity 97%
Chemical Name or Material Ethyl 3-oxocyclobutanecarboxylate
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.