accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NHSOOBr

CAS RN 871269-07-7

IUPAC Name: 3-bromo-N-ethylbenzene-1-sulfonamide
Synonyms: 3-bromo-N-ethylbenzenesulfonamide
Molecular Weight (g/mol): 264.14
Molecular Formula: C8H10BrNO2S
InChi Key: MADIDEQQKAOBFI-UHFFFAOYSA-N
SMILES: CCNS(=O)(=O)C1=CC(Br)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results