accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 871329-64-5

IUPAC Name: (5-cyano-1H-indol-2-yl)boronic acid
Synonyms: 5-cyano-1H-indol-2-ylboronic acid
Molecular Weight (g/mol): 185.98
Molecular Formula: C9H7BN2O2
InChi Key: UALSJXICMYXRMN-UHFFFAOYSA-N
SMILES: OB(O)C1=CC2=CC(=CC=C2N1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results