Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Cyanoindole-2-boronic acid, Thermo Scientific™

Catalog No. AAH53203MD
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH53203MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH53203MD Supplier Thermo Scientific Chemicals Supplier No. H53203MD

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 871329-64-5
Molecular Formula C9H7BN2O2
Molecular Weight (g/mol) 185.98
MDL Number MFCD06798223
InChI Key UALSJXICMYXRMN-UHFFFAOYSA-N
Synonym 5-cyano-1h-indole-2-boronic acid, 5-cyanoindole-2-boronic acid, 5-cyano-1h-indol-2-yl boronic acid, boronic acid,b-5-cyano-1h-indol-2-yl, acmc-209qg8, 5-cyano-2-indoleboronic acid, boronic acid, 5-cyano-1h-indol-2-yl
PubChem CID 25067270
IUPAC Name (5-cyano-1H-indol-2-yl)boronic acid
SMILES OB(O)C1=CC2=CC(=CC=C2N1)C#N

Specifications

Melting Point 220°C to 226°C
Quantity 250 mg
Formula Weight 185.98
Chemical Name or Material 5-Cyanoindole-2-boronic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.