accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOCH3FFFHOOH

CAS RN 871329-83-8

IUPAC Name: [4-ethoxy-3-(trifluoromethyl)phenyl]boronic acid
Synonyms: 4-ethoxy-3-(trifluoromethyl)phenylboronic acid
Molecular Weight (g/mol): 233.98
Molecular Formula: C9H10BF3O3
InChi Key: SHCPXHBLEGXHIV-UHFFFAOYSA-N
SMILES: CCOC1=C(C=C(C=C1)B(O)O)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results