CAS RN 871332-98-8
IUPAC Name:
(5-bromo-2-ethoxypyridin-3-yl)boronic acid
Synonyms:
5-bromo-2-ethoxypyridin-3-ylboronic acid
Molecular Weight (g/mol):
245.87
Molecular Formula:
C7H9BBrNO3
InChi Key:
VXXXKWUXSALQQW-UHFFFAOYSA-N
SMILES:
CCOC1=NC=C(Br)C=C1B(O)O
1
–
1
of
1
results
1
5-Bromo-2-ethoxypyridine-3-boronic acid, 97%, Thermo Scientific™
CAS: 871332-98-8 Molecular Formula: C7H9BBrNO3 Molecular Weight (g/mol): 245.87 MDL Number: MFCD07363847 InChI Key: VXXXKWUXSALQQW-UHFFFAOYSA-N Synonym: 5-bromo-2-ethoxypyridine-3-boronic acid,5-bromo-2-ethoxypyridin-3-yl boronic acid,5-bromo-2-ethoxy-3-pyridyl boronic acid,5-bromo-2-ethoxy-3-pyridineboronic acid,boronic acid,b-5-bromo-2-ethoxy-3-pyridinyl,acmc-209qhr,5-bromo-2-ethoxypyridin-3-ylboronicacid,abbypharma ap-12-5495,5-bromo-2-ethoxy-3-pyridinyl boronic acid,5-bromanyl-2-ethoxy-pyridin-3-yl boronic acid PubChem CID: 44886948 IUPAC Name: (5-bromo-2-ethoxypyridin-3-yl)boronic acid SMILES: CCOC1=NC=C(Br)C=C1B(O)O