accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 871333-99-2

IUPAC Name: {1,7,7-trimethylbicyclo[2.2.1]hept-2-en-2-yl}boronic acid
Synonyms: 1,7,7-trimethylbicyclo[2.2.1]hept-2-en-2-ylboronic acid
Molecular Weight (g/mol): 180.05
Molecular Formula: C10H17BO2
InChi Key: ZGDNWNBGFZOLLT-UHFFFAOYNA-N
SMILES: CC1(C)C2CCC1(C)C(=C2)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results