Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1S)-1,7,7-Trimethylnorborn-2-ene-2-boronic acid, 95%, Thermo Scientific™

Catalog No. AAH52956MD
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52956MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH52956MD Supplier Thermo Scientific Chemicals Supplier No. H52956MD

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 871333-99-2
Molecular Formula C10H17BO2
Molecular Weight (g/mol) 180.05
MDL Number MFCD08705231
InChI Key ZGDNWNBGFZOLLT-UHFFFAOYNA-N
Synonym 1s-1,7,7-trimethylbicyclo 2.2.1 hept-2-en-2-ylboronic acid, 1s-1,7,7-trimethylbicyclo 2.2.1 hept-2-en-2-yl boronic acid, boronic acid,b-1s,4r-1,7,7-trimethylbicyclo 2.2.1 hept-2-en-2-yl, 1s-1,7,7-trimethyl bicyclo 2.2.1 hept-2-en-2-ylboronic acid
PubChem CID 44119811
IUPAC Name [(4S)-4,7,7-trimethyl-3-bicyclo[2.2.1]hept-2-enyl]boronic acid
SMILES CC1(C)C2CCC1(C)C(=C2)B(O)O

Specifications

Melting Point 205°C to 207°C
Quantity 250 mg
Formula Weight 180.05
Percent Purity 95%
Chemical Name or Material (1S)-1,7,7-Trimethylnorborn-2-ene-2-boronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.