accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3HNON

CAS RN 871825-60-4

IUPAC Name: methyl({[3-(pyridin-2-yloxy)phenyl]methyl})amine
Synonyms: methyl({[3-(pyridin-2-yloxy)phenyl]methyl})amine
Molecular Weight (g/mol): 214.27
Molecular Formula: C13H14N2O
InChi Key: OEPWSCXLHMBFPC-UHFFFAOYSA-N
SMILES: CNCC1=CC(OC2=CC=CC=N2)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results