accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SOO

CAS RN 872-93-5

IUPAC Name: 3-methyl-1λ⁶-thiolane-1,1-dione
Synonyms: 3-methyl sulfolane
Molecular Weight (g/mol): 134.19
Molecular Formula: C5H10O2S
InChi Key: CMJLMPKFQPJDKP-UHFFFAOYNA-N
SMILES: CC1CCS(=O)(=O)C1
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)