Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methylsulfolane, MP Biomedicals

Catalog No. ICN216117 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN216117 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN216117 Supplier MP Biomedicals, Inc Supplier No. 0521611710

Chemical Identifiers

CAS 872-93-5
Molecular Formula C5H10O2S
Molecular Weight (g/mol) 134.193
InChI Key CMJLMPKFQPJDKP-UHFFFAOYSA-N
Synonym 3-methylsulfolane, 3-methyl sulfolane, 3-methyltetrahydrothiophene 1,1-dioxide, thiophene, tetrahydro-3-methyl-, 1,1-dioxide, acmc-209qjh, 3-methylthiolane-1,1-dione, 3-methylthiacyclopentane 1,1-dioxide, 3-methyl-2,4-dihydrothiophene 1,1-dioxide, thiophene,tetrahydro-3-methyl-, 1,1-dioxide, 1,1-dioxo-1??-thiolan-3-yl methyl
PubChem CID 79119
IUPAC Name 3-methylthiolane 1,1-dioxide
SMILES CC1CCS(=O)(=O)C1

Specifications

Quantity 10 g
Formula Weight 134.19
Chemical Name or Material 3-Methylsulfolane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.