accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOHHO

CAS RN 872050-52-7

IUPAC Name: [4-(pyren-1-yl)phenyl]boronic acid
Synonyms: 4-(pyren-1-yl)phenylboronic acid
Molecular Weight (g/mol): 322.17
Molecular Formula: C22H15BO2
InChi Key: ISMRPUXCQLIMJL-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C(C=C1)C1=CC=C2C=CC3=CC=CC4=CC=C1C2=C34
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results