Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(1-Pyrenyl)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. P23055G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P2305-5G 5 g
P23051G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P2305-5G Supplier TCI America Supplier No. P23055G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 872050-52-7
Molecular Formula C22H15BO2
Quantity 5 g
MDL Number MFCD28130386
Synonym 4-(1-Pyrenyl)benzeneboronic Acid
InChI Key ISMRPUXCQLIMJL-UHFFFAOYSA-N
SMILES B(C1=CC=C(C=C1)C2=C3C=CC4=CC=CC5=C4C3=C(C=C5)C=C2)(O)O
IUPAC Name (4-pyren-1-ylphenyl)boronic acid
Molecular Weight (g/mol) 322.17
PubChem CID 59217173
Formula Weight 322.17
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.